Learn More
nucleobindin 1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 164681AP
Catalog No. 50-173-0965
Add to cart
Description
Description
Nucleobindin 1 encodes a member of a small calcium-binding EF-hand protein family. The encoded protein is thought to have a key role in Golgi calcium homeostasis and Ca(2+)-regulated signal transduction events.
Specifications
Specifications
nucleobindin 1
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q02818, Q02819
CALNUC, NUC, NUCB1, nucleobindin 1
Rabbit
IgG
150 μL
-20°C
Primary
18220, 4924
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NUCB1
Liquid
Nucb1
nucleobindin 1 Fusion Protein Ag9548
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse