Learn More
Abnova™ Nucleohistone (Calf)
Used for SDS-PAGE
Manufacturer:  Abnova™ P5240
Catalog No. 89-954-777
Add to cart
Description
Description
#REF!
Specifications
Specifications
A complex of histone and DNA prepared from Calf Thymus according to Zamenhof, S. Methods in Enzymology, 3, 696, (1957)
Lyophilized
A complex of histone and DNA prepared from Calf Thymus according to Zamenhof, S. Methods in Enzymology, 3, 696, (1957)
Store at 4°C.
NUCLEOHISTONE
None
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Nucleohistone (Calf)
250 mg
RUO
Bovine