Learn More
Enzo Life SciencesSupplier Diversity Partner Nucleolin monoclonal antibody (4E2) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIKAMCP100E
Clone: 4E2. Host: Mouse. Isotype: IgG1. Immunogen: Human nucleolin from Raji cell extract. UniProt: P19338. Species Reactivity: Human. Applications: Flow Cytometry, ICC, IHC, WB. Recommended dilutions: Flow Cytometry (5-10µg/ml final. Concentration). Immunocytochemistry (1-10µg/ml). Immunohistochemistry (1-10µg/ml). Western Blot (1µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-666-473