Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleolin Mouse anti-Canine, Human, Active Motif™ Non-distribution product as customer accommodation.

Mouse Recombinant Monoclonal Antibody

$98.40 - $465.85

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen Nucleolin
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3762 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
91232
10 μg
Each for $98.40
Add to cart
 
50-199-3761 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
91231
100 μg
Each for $465.85
Add to cart
 
Description

Description

Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20.
Specifications

Specifications

Monoclonal
Primary
Nucleolin
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit