Learn More
GE Healthcare Nucleon™ BACC1 Genomic DNA Extraction Kit PROMO
Nucleon BACC DNA Extraction kits use a Nucleon resin to bind proteins ensuring high yields of high-quality, high molecular weight genomic DNA in only one hour
Manufacturer:  GE Healthcare 29361648
Catalog No. 45-000-955PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Phenol-free protocol requiring only 30 minutes to complete
  • Low-shearing protocol gives maximum recoveries of high-molecular weight DNA
  • For 50 preps of 1mL of whole blood or cultured cells (1 to 3 x 106 cells/mL)
  • Available in different formats/reaction scales (See 45-000-956 and 45-000-960)
Specifications
Specifications
illustra Nucleon BACC system rapidly extracts high molecular weight DNA from whole blood and cultured cells.
Reagents A and B, 5M Sodium Perchlorate and Nucleon™ Resins
25 kits
< 12 μg DNA/mg tissue
Rapid extraction of high-quality, high molecular weight genomic DNA from cell cultures and blood.
illustra Nucleon BACC1
Cell cultures and blood