Learn More
GE Healthcare Nucleon™ BACC2 Genomic DNA Extraction Kit : PROMOTION
Promotion Details: Receive a 31% discount.
Kits use a Nucleon resin to bind proteins ensuring high yields of high-quality, high molecular weight genomic DNA in only one hour
Manufacturer:  GE Healthcare 29361649
Catalog No. 45-000-956PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Phenol-free protocol requiring only 30 minutes to complete
  • Low-shearing protocol gives maximum recoveries of high-molecular weight DNA
  • For 50 preps of 10mL of whole blood or cultured cells (3 to 10 x 106 cells/10mL), or 100 preps of 1mL of whole blood or cultured cells (1 to 3 x 106 cells/mL))
  • Available in different formats/reaction scales (see 45-000-955 and 45-000-960)
Additional Reagent A will need to be prepared to process all 50 x 10mL preps
Specifications
Specifications
Rapid extraction of high-quality, high-molecular weight genomic DNA from blood and cell cultures
Reagents A and B, 5M Sodium Perchlorate and Nucleon™ Resins
Nucleon BACC2 Genomic DNA Extraction Kit
50 Preps