Learn More
GE Healthcare Nucleon™ HT Genomic DNA Extraction Kit : PROMOTION
Promotion Details: Receive a 31% discount.
Phenol-free extraction of genomic DNA from paraffin-embedded sections and hard tissue requiring proteinase K digestion. Consistent extraction of high-quality, amplifiable DNA. Protocols developed for those tissues which don't homogenize easily in lysis bu
Manufacturer:  GE Healthcare 29361650
Catalog No. 45-000-957PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Extract DNA from FFPE samples, mouse tail, Xiphisternum etc.
  • User-friendly protocol enables efficient processing of large numbers of samples
  • Patented Nucleon resin delivers highly purity by binding macromolecules other than DNA
  • DNA is not bound to any solid surface throughout the purification avoiding exposure to shearing forces promoting recovery of high yields of highly intact DNA
  • Each kit is designed to recover high-quality DNA from 50 preparations of up to 25mg of hard tissue or 50 paraffin sections
Specifications
Specifications
Extract genomic DNA from Paraffin embedded sections and hard tissue requiring Proteinase K digestion
18mL Reagent B, 6mL 5M Sodium Perchlorate, 8mL Nucleon™ Resin, 10mg Proteinase K
50 Preps