Learn More
GE Healthcare Nucleon™ PhytoPure™ Genomic DNA Extraction Kits: PROMO
Product
Manufacturer:  GE Healthcare RPN8510PM
Catalog No. 45-000-958LR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Includes: Reagents 1 and 2 and PhytoPure Resin
Description
Description
  • Rapid protocol does not require phenol or CTAB
  • Two kit sizes provide economical scale-up using the same phenol-free protocol
  • Effective removal of polysaccharides minimises inhibition of downstream applications
Specifications
Specifications
High yields of high-quality plant genomic DNA
Reagents 1 and 2 and PhytoPure™ Resin
Kit
100mg