Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Nucleophosmin (NPM) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Nucleophosmin (NPM) Ab-1, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS1849R1

 View more versions of this product

Catalog No. MS1849R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Nucleophosmin (NPM)/B23/numatrin is a phosphoprotein associated with cell nucleoli. Treatment of cells with cytotoxic drugs may make nucleophosmin enter nucleoplasm. It is reported that nucleophosmin shuttles continuously between cytoplasm and nucleoli to carry newly synthesized ribosomal proteins into nucleoli. NPM-ALK is a hybrid of the anaplastic lymphoma kinase (ALK) gene and the nucleophosmin (NPM) gene resulting from the t(2;5)(p23;q35) translocation found in anaplastic large cell lymphoma. In acute promyelocytic leukemia NPM gene is fused to RARalpha gene in (5;17) (q33;q12) transition.

Host Species: Mouse

Clone: NA24

Isotype: IgG/κ

Species Reactivity: Human. Others not known.

Epitope: N-terminal

Immunogen: GST fusion protein containing the N-terminal part of nucleophosmin fused to 14aa of ALK protein

Molecular Weight: 38kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Nucleophosmin (NPM) Ab-1
Monoclonal
Unconjugated
Mouse
1 mL
Cancer and Tumor Biology
Human
IgG κ
Immunohistochemistry (Paraffin)
NA24
Ready-to-use for IHC (UVLP)
GST fusion protein containing the N-terminal part of nucleophosmin fused to 14 aa of ALK protein
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit