Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleoporin 107 Rabbit anti-Human, DyLight 755, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299074IR
Description
Nucleoporin 107 Polyclonal specifically detects Nucleoporin 107 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Nucleoporin 107 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
DyLight 755 | |
nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Nucleoporin 107. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
57122 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title