Learn More
Nucleostemin Mouse anti-Human, Mouse, HRP, Clone: 2C8D5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237410H
Catalog No. NBP237410H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Nucleostemin
2C8D5
Western Blot, ELISA
Purified
GNL3
Purified recombinant fragment of human GNL3 (AA: 1-226) expressed in E. Coli.
Protein A or G purified
Apoptosis, Cellular Markers, Chromatin Modifiers, Core ESC Like Genes, Embryonic Stem Cell Markers, Epigenetics, Histones and Modified Histones, Metabotropic Glutamate Receptors, microRNA, mTOR Pathway, Neurofilaments, Stem Cell Markers, Stem Cell Signaling Pathway
Primary
26354
ELISA, Western Blot
HRP
PBS with No Preservative
C77032, E2IG3NNP47, estradiol-induced nucleotide binding protein, guanine nucleotide binding protein-like 3 (nucleolar), guanine nucleotide-binding protein-like 3, MGC800, Novel nucleolar protein 47, NSE2-induced gene 3 protein, Nucleolar GTP-binding protein 3, nucleostemin
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse