Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleotide binding protein like Mouse anti-Human, Mouse, Rat, DyLight 550, Clone: OTI5D5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273094R
Description
Nucleotide binding protein like Monoclonal specifically detects Nucleotide binding protein like in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Nucleotide binding protein like | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NUBPL | |
Recombinant protein fragment corresponding to amino acids 1-250 of human Nucleotide binding protein like (NP_079428) produced in HEK293T cells. | |
0.1 mL | |
80224.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5D5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
C14orf127, FLJ12660, huInd1IND1 homolog, IND1chromosome 14 open reading frame 127, iron-sulfur protein NUBPL, iron-sulfur protein required for NADH dehydrogenase, nucleotide binding protein-like, Nucleotide-binding protein-like | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title