Learn More
Nucleotides Model Set Products for Science Education
Illustrate the structure of nucleotides with this model.
Manufacturer:  Ryler Enterprises NUC2
Catalog No. S94031
Add to cart
Includes: Kit Contents:
  • 60 white 1-peg hydrogen atom centers
  • 16 blue 4-peg nitrogen atom centers
  • 5 red 4-peg oxygen atom centers
  • 25 red 2-peg oxygen atom centers
  • 40 black 4-peg carbon atom centers
  • 3 purple 5-peg phosphorous atom centers
  • 128-1.25 in. PVC, clear bonds (single bonds)
  • 30-1.625 in. PVC, clear bonds (double bonds)
  • 6-2 in. PVC, white bonds (H bonds)
  • 5-.75 in. PVC, white bonds (glycosidic bonds)
  • Lab procedure
  • Answer key
Description
Description
  • Color-coded, modeled plastic atom centers are 1/4 in. diameter
  • Pegs are 3/8 in. long and are set at angles peculiar to the elements the atoms represent
  • Standard color-coding
  • Atom centers are bonded by connecting pegs with pvc tubing
Specifications
Specifications
Build an H bond Adenine to Thymine and Guanine to Cytosine
60 White 1-peg Hydrogen atom centers, 16 blue 4-peg Nitrogen atom centers, 5 red 4-peg Oxygen atom centers, 25 red 2-peg Oxygen atom centers, 40 black 4-peg Carbon atom centers, 3 purple 5-peg Phosphorous atom centers, One hundred twenty eight 1.25 in. PVC, clear bonds (single bonds), Thirty 1.625 in. PVC, clear bonds (double bonds), Six 2 in. PVC, white bonds (H bonds), Five 0.75 in. PVC, white bonds (Glycosidic bonds), lab procedure, answer key
Nucleotides model set