Learn More
GE Healthcare Nuclepore Polycarbonate Hydrophilic Membranes
Captures extremely small particles.
$168.00 - $688.50
Specifications
Hydrophobicity Hydrophilic
For Use With (Application) For filtering small particulates
Material Nuclepore polycarbonate
View More Specs
Products 2
Catalog Number Mfr. No. Pore Size Format Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Format Dimensions (L x W) Price Quantity & Availability  
09-940--071 View Documents GE Healthcare
113506
0.2μm Sheet 20.3 x 25.4cm Pack of 25 for $688.50
Add to cart
09-940--072 View Documents GE Healthcare
111207
0.4μm Circle 0.5μm Pack of 100 for $168.00
Add to cart
Description
Description
  • Track-etched for highly defined pore size, high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance
  • Good thermal stability for wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
Specifications
Specifications
Hydrophilic
For filtering small particulates
Nuclepore polycarbonate