Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDCD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554012
Description
Immunogen sequence: SKVGEYWWNA ILEGEEPIDI DKINKERSMA TVDEEEQAVL DRLTFDYHQK LQGKPQSHEL KVHEMLKKGW DAEGSPFRGQ RFDPAMF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 27%.
NUDCD3 gene ontology annotations related to this gene include cilium assembly; developmental process; protein folding; protein localization to pericentriolar material.
Specifications
NUDCD3 | |
Polyclonal | |
Unconjugated | |
NUDCD3 | |
AI427847; BC024322; KIAA1068; KIAA1068 protein; mKIAA1068; nudC domain containing 3; nudC domain-containing protein 3; nudcd3; NudCL; NudC-like protein; Unknown (protein for MGC:137347); zgc:77067 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23386 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVD9 | |
NUDCD3 | |
Recombinant protein corresponding to Human NUDCD3. Recombinant protein control fragment (Product #RP-92612). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction