Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDCD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554013
Description
Immunogen sequence: NPDSYNGAVR ENYTWSQDYT DLEVRVPVPK HVVKGKQVSV ALSSSSIRVA MLEENGERVL MEGKLTHKIN TESSLWSL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 30%.
NUDCD3 gene ontology annotations related to this gene include cilium assembly; developmental process; protein folding; protein localization to pericentriolar material.
Specifications
NUDCD3 | |
Polyclonal | |
Unconjugated | |
NUDCD3 | |
AI427847; BC024322; KIAA1068; KIAA1068 protein; mKIAA1068; nudC domain containing 3; nudC domain-containing protein 3; nudcd3; NudCL; NudC-like protein; Unknown (protein for MGC:137347); zgc:77067 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100125378, 23386 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVD9 | |
NUDCD3 | |
Recombinant protein corresponding to Human NUDCD3. Recombinant protein control fragment (Product #RP-92611). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction