Learn More
NUDT12 Mouse anti-Human, Clone: OTI1A2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805547
Catalog No. 50-166-7474
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.
Specifications
Specifications
NUDT12
Monoclonal
Unconjugated
NUDT12
Lyophilized
NUDT12
Human recombit protein fragment corresponding to amino acids 1-251 of human NUDT12 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1A2
PBS with 8% trehalose and no preservative; pH 7.3
Q9BQG2
nudix hydrolase 12
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
83594