Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT12 Mouse anti-Human, Clone: OTI2G2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526745

Catalog No. PIMA526745


Add to cart

Description

Description

Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances (Abdelraheim et al., 2003
Specifications

Specifications

NUDT12
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BQG2
nucleoside diphosphate linked moiety X-type motif 12, nucleoside diphosphate-linked moiety X motif 12, nudix motif 12, peroxisomal NADH pyrophosphatase NUDT12
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83594
Immunohistochemistry (Paraffin), Western Blot
OTI2G2
Unconjugated
NUDT12
Liquid
NUDT12
Human recombit protein fragment corresponding to amino acids 1-251 of NUDT12 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit