Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT19 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548218

Catalog No. PA548218

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NUDT19 is a coenzyme A diphosphatase that mediates the hydrolysis of a wide range of CoA esters, including choloyl-CoA and branched-chain fatty-acyl-CoA esters. At low substrate concentrations medium and long-chain fatty-acyl-CoA esters, NUDT19 are the primary substrates.
Specifications

Specifications

NUDT19
Polyclonal
PBS with 0.09% sodium azide
A8MXV4
Nucleoside diphosphate-linked moiety X motif 19, mitochondrial, Nudix motif 19, 361-, NUDT19
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
390916
Flow Cytometry, Western Blot
Unconjugated
NUDT19
Liquid
NUDT19
KLH conjugated synthetic peptide between 153-181 amino acids from the Central region of human NUDT19.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit