Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT21 Mouse anti-Human, Clone: OTI5B3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526808

Catalog No. PIMA526808


Add to cart

Description

Description

NUDT21 is one subunit of a cleavage factor required for 3 ft. RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3 ft. end processing complex and facilitates the recruitment of other processing factors.The protein encoded by this gene is one subunit of a cleavage factor required for 3 ft. RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3 ft. end processing complex and facilitates the recruitment of other processing factors. This gene encodes the 25kD subunit of the protein complex, which is composed of four polypeptides.
Specifications

Specifications

NUDT21
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O43809
Nudix (nucleoside diphosphate linked moiety X)-type motif 21, CPSF5, CFIM25
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11051
Immunohistochemistry (Paraffin), Western Blot
OTI5B3
Unconjugated
NUDT21
Liquid
NUDT21
Human recombit protein fragment corresponding to amino acids 2-227 of NUDT21 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit