Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT22 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551214

Catalog No. PA551214


Add to cart

Description

Description

The antibody detects endogenous levels of total NUDT22 protein.

NUDT22 (nudix (nucleoside diphosphate linked moiety X)-type motif 22) is a member of the Nudix hydrolase family of pyrophosphatases. Nudix hydrolases contain a characteristic Nudix domain and are responsible for catalyzing the hydrolysis of nucleoside diphosphate derivatives. NUDT22 is a 303 amino acid protein that lacks the nudix box, therefore lacking hydrolase activity. The gene encoding NUDT17 maps to human chromosome 11, which makes up around 4% of human genomic DNA and is considered a gene and disease association dense chromosome.
Specifications

Specifications

NUDT22
Polyclonal
Unconjugated
NUDT22
Liquid
NUDT22
Fusion protein of human NUDT22
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9BRQ3
MGC13045, Nucleoside diphosphate-linked moiety X motif 22, NUD22
Rabbit
IgG
100 μL
-20°C
Primary
84304
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit