Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548825

Catalog No. PA548825

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with rat based on sequence homology.

Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), PP-InsP4 and [PP]2-InsP4 (bisdiphosphoinositol tetrakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphate Ap6A, but not Ap5A. The major reaction products are ADP and p4a from Ap6A. Also able to hydrolyze 5-phosphoribose 1-diphosphate. Does not play a role in U8 snoRNA decapping activity. Binds U8 snoRNA.
Specifications

Specifications

NUDT4
Polyclonal
PBS with 0.09% sodium azide
Q9NZJ9, Q8R2U6
Diphosphoinositol polyphosphate phosphohydrolase 2, DIPP-2, Diadenosine 5′, 5′′′-P1, P6-hexaphosphate hydrolase 2, 361-, Nucleoside diphosphate-linked moiety X motif 4, Nudix motif 4, NUDT4, DIPP2, KIAA0487
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11163, 71207
Western Blot
Unconjugated
NUDT4
Liquid
NUDT4
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human NUDT4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit