Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT6 Mouse anti-Human, Clone: OTI7F4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525396

Catalog No. PIMA525396


Add to cart

Description

Description

NUDT6 (nudix (nucleoside diphosphate linked moiety X)-type motif 6), also known as Antisense basic fibroblast growth factor or GFG-1, is a member of the Nudix hydrolase family of pyrophosphatases. Nudix hydrolases contain a characteristic Nudix domain and are responsible for catalyzing the hydrolysis of nucleoside diphosphate derivatives. The gene encoding NUDT6 is a FGF-2 gene antisense transcript, and NUDT6 is believed to regulate FGF-2 expression. FGF-2 is a mutifunctional heparin-binding growth factor important to angiogenesis, neuroectoderm development and wound healing. NUDT6 is expressed as two isoforms produced by alternative splicing.
Specifications

Specifications

NUDT6
Monoclonal
0.63 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P53370
Antisense basic fibroblast growth factor, FGF2AS, GFG1, Nucleoside diphosphate linked moiety X motif 6, Nucleoside diphosphate-linked moiety X motif 6, Nudix motif 6, NUDT6, NUDT6, Protein GFG
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11162
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI7F4
Unconjugated
NUDT6
Liquid
NUDT6
Full length human recombit protein of NUDT6 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit