Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuf2 Mouse anti-Human, DyLight 755, Clone: 644, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243779IR

Catalog No. NBP243779IR


Add to cart

Description

Description

Nuf2 Monoclonal antibody specifically detects Nuf2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Nuf2
644
Western Blot
Purified
NUF2
The exact sequence is proprietary.
Protein A purified
Cell Cycle and Replication
Primary
83540
Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit