Alfa Aesar™ Nujol (light oil)

Manufacturer: Alfa Aesar™ H316610F

Quantity 2500mL

 View more versions of this product

Catalog No. AAH316610F

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 8012-95-1
    MDL Number MFCD00131611
    Synonym Mineral oil; Paraffin oil
    Chemical Name or Material Nujol (light oil)

    Specifications

    TSCA TSCA
    MDL Number MFCD00131611
    CAS 8012-95-1
    RTECS Number PY8030000
    Flash Point 220°C (428°F)
    Boiling Point >300°C
    Melting Point ≈−20°C
    Quantity 2500mL
    Synonym Mineral oil; Paraffin oil
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Chemical Name or Material Nujol (light oil)

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit