Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nujol (light oil)

Click to view available options
Quantity:
50mL
500mL
2500mL
8012-95-1
C15H11ClO7
338.696
MFCD00131611
FFNDMZIBVDSQFI-UHFFFAOYSA-N
delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
68245
CHEBI:38701
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
C1=C(C=C(C(=C1O)O)O)C2=C(C=C3C(=CC(=CC3=[O+]2)O)O)O.[Cl-]
8012-95-1
C15H11ClO7
338.696
MFCD00131611
FFNDMZIBVDSQFI-UHFFFAOYSA-N
delphinidin chloride,delphinidin,delphinidine,delphinidol,ephdine,delfinidol chloride,unii-em6md4aehe,ccris 2518,em6md4aehe,3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
68245
CHEBI:38701
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
C1=C(C=C(C(=C1O)O)O)C2=C(C=C3C(=CC(=CC3=[O+]2)O)O)O.[Cl-]
Flash Point 220°C (428°F)
Boiling Point >300°C
Density 0.838
Refractive Index 1.4613
Quantity 50mL
Melting Point ~-20°C
Chemical Name or Material Nujol (light oil)

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 232-384-2

RTECSNumber : PY8030000

TSCA : Yes

Recommended Storage : Ambient temperatures