Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NULP1 Mouse anti-Human, Clone: PCRP-TCF25-1A11, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen NULP1
Clone PCRP-TCF25-1A11
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array
Applications Western Blot, Flow Cytometry, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122192
View Documents
Novus Biologicals
NBP30735920UG
20 μg
Each for $138.37
Add to cart
 
NB122191
View Documents
Novus Biologicals
NBP30735910UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

NULP1 Monoclonal antibody specifically detects NULP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array
Specifications

Specifications

NULP1
0.2 mg/ml
Western Blot, Flow Cytometry, Peptide Array
Purified
RUO
10 mM PBS with 0.05% BSA
22980
IgG2a
Primary
Store at 4°C.
PCRP-TCF25-1A11
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array
Unconjugated
Mouse
Human
hKIAA1049, KIAA1049Hulp1, Nuclear localized protein 1, Nulp1, NULP1PRO2620, TCF-25, transcription factor 25, transcription factor 25 (basic helix-loop-helix)
Recombinant full-length human NULP1 protein
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit