Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ NuMA Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

$230.50

Specifications

Product Type Recombinant Protein Antigen
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol NUMA1
View More Specs

Products 2
Catalog Number Mfr. No. Protein Length Price Quantity & Availability  
Catalog Number Mfr. No. Protein Length Price Quantity & Availability  
NBP254672X View Documents Novus Biologicals™
NBP254672PEP
IRFLELQKVASSSSGNNFLSGSPASPMGDILQTPQFQMRRLKKQLADERSNRDELELELAENRKLLTEKDAQIAMMQQRIDRLALLNEKQAASPLEPKEL Each for $230.50
Add to cart
 
NBP254673X View Documents Novus Biologicals™
NBP254673PEP
TEEGQQILKQPVSERLDFVCSFLQKNRKHPSSPECLVSAQKVLEGSELELAKMTMLLLYHSTMSSKSPRDWEQFEYKIQAELAVILKFVLDHEDGLNLNEDLENFLQKAPVPSTCSST Each for $230.50
Add to cart
 
Description

Description

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human NuMA The NuMA Recombinant Protein Antigen is derived from E. coli. The NuMA Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications

Specifications

Recombinant Protein Antigen
RUO
NUMA1
NUMA1 Recombinant Protein Antigen
PBS and 1M Urea, pH 7.4
E.coli
Unlabeled
Yes
Store at −20°C long term. Avoid freeze-thaw cycles.
centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic, apparatus protein 1, NuMA protein
>80% by SDS-PAGE and Coomassie blue staining
Human
AC

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit