Learn More
Aviva Systems Biology Corp NUMA1 Antibody - C-terminal region (ARP50532_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUMA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NUMA1. Alternate names: NUMAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60367