Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Numb Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: OTI4F3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273129IR
Description
Numb Monoclonal specifically detects Numb in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Numb | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NUMB | |
Full length recombinant protein of human Numb (NP_003735) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI4F3 | |
Western Blot, Immunohistochemistry | |
h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
Mouse | |
Affinity Purified | |
Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
8650.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title