Learn More
Numb Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF4338SP
Catalog No. AF4338SP
Add to cart
This item is not returnable. View return policy
Description
Description
Numb Polyclonal antibody AF4338SP specifically detects Numb in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
Numb
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
AAH68476
h-Numb, mnb, m-numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Numb in direct ELISAs and Western blots.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NUMB
E. coli-derived recombinant human Numb Thr356-Leu592 Accession # AAH68476
Antigen Affinity-purified
RUO
Primary
8650
Reconstitute at 0.2 mg/mL in sterile PBS.