Learn More
Thermo Scientific™ Nunc™ 10/11mL Polystyrene Centrifuge Tubes
Increase the effectiveness of centrifugation with these polycarbonate centrifuge tubes, which have been both RCF and tightness tested.
Specifications
Certifications/Compliance CE marked
Material Polystyrene
Max. Force 3,000 x g
No. per Case 480
View More Specs
Products 1
Catalog Number Mfr. No. Bottom Shape Graduations Writing Area Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Bottom Shape Graduations Writing Area Sterility Price Quantity & Availability  
12-565-190
View Documents
Thermo Scientific™
362707
Flat None Yes Sterile This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Increase the effectiveness of centrifugation with Thermo Scientific™ Nunc™ 10/11mL Polystyrene Centrifuge Tubes, which have been both RCF and tightness tested.
  • Made in a variety of formats for greater flexibility
  • Different bottom shapes to meet all needs
  • Include polyethylene plug seal screw caps
  • Fit most standard centrifuges
  • Relative centrifugal force tested (RCF)
  • CE marked
  • Specifications
    Specifications
    CE marked
    Polystyrene
    3,000 x g
    480