Learn More
Thermo Scientific™ Nunc™ 1536 Well High Base Microplates Available on GSA/VA Contract for Federal Government customers only.
Work with small volumes and screen 4X as many samples in one plate. Thermo Scientific™ Nunc™ 1536 Well High Base Microplates are ideal for high-throughput screening.
Manufacturer:  Thermo Scientific™ 164708
 View more versions of this product
Catalog No. 12-565-499
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Work with small volumes and screen 4X as many samples in one plate. Thermo Scientific™ Nunc™ 1536 Well High Base Microplates are ideal for high-throughput screening; the skirted design around the base of the plate is optimal for robotic handling.
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth.
  • Polystyrene
  • Working range: 2–10µL/well
  • Customized bar coding is available
Description & Specifications
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
1536 Well High Base Plate, Polysytrene; Cell Culture, Black, with lid, Sterile
25
Cell Culture
1536 F
Black
100
Yes
13.4µL