Thermo Scientific™ Nunc™ 1536 Well High Base Microplates Available on GSA/VA Contract for Federal Government customers only.
Work with small volumes and screen 4X as many samples in one plate. Thermo Scientific™ Nunc™ 1536 Well High Base Microplates are ideal for high-throughput screening.
Manufacturer: Thermo Scientific™ 164707
 View more versions of this product
Catalog No. 12-565-498
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Description 1536 Well High Base Plate, Polysytrene; Cell Culture, Clear, with lid, Sterile
    No. per Case 100
    No. per Pack 25,25
    Sterile Yes
    Surface Coating Cell Culture
    Volume (Metric) Total 13.4µL,13.4µL
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Well Design 1536 F
    Color Clear
    Item Description Cell Culture, Clear, with lid, Sterile
    Work with small volumes and screen 4X as many samples in one plate. Thermo Scientific™ Nunc™ 1536 Well High Base Microplates are ideal for high-throughput screening; the skirted design around the base of the plate is optimal for robotic handling.
    • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
    • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
    • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth.
    • Polystyrene
    • Working range: 2–10µL/well
    • Customized bar coding is available
  • Description & Specifications
Related Products
Product Content Feedback