Learn More
Thermo Scientific™ Nunc™ 1536-Well, Non-Treated, Flat-Bottom Microplate
Color:White; Sterility:Non-sterile; Lid:Without Lid
Manufacturer:  Thermo Scientific™ 264712
Catalog No. 12-565-490
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Work with small volumes and screen 4X as many samples in one plate. Thermo Scientific™ Nunc™ 1536 Well High Base Microplates are ideal for high-throughput screening; the skirted design around the base of the plate is optimal for robotic handling.
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth.
  • Polystyrene
  • Working range: 2–10μL/well
  • Customized bar coding is available
Specifications
Specifications
Flat
White
Polystyrene
25
Non-sterile
1536
Nunc™
Skirted
10 mL
Microplate
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
1536 Well High Base Plate, Polysytrene; Non-Treated, White, without lid, Non-sterile
100
13.4 μL
Non-treated
32 x 48
Without Lid
13.4 µL
25/Pk., 100/Cs.
Standard