Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates Available on GSA/VA Contract for Federal Government customers only.
Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates.
Manufacturer: Thermo Scientific™ 267459
 View more versions of this product
Catalog No. 12-566-210
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Lid No,No
    No. of Wells 384
    No. per Case 100
    No. per Pack 25,25
    Sterile No
    Surface Coating Non-Treated
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Volume (Metric) Working 2 to 35µL
    Well Design Square Rounded
    Closure No
    Color Natural
    Wells 384
    Well Shape Square Rounded
    Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors to allow for quick identification of storage plates.
    Features:
    • Rounded wells are optimized for mixing and sample recovery
    • Square rounded wells minimize wicking (capillary action)
    • Natural plates for storage identification or storing colorimetric assays
    • White plates for luminescent assays
    • Black plates for fluorescent assays
    • Customized bar coding is available
  • Description & Specifications
Related Products
Product Content Feedback