Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Storage Microplates

Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates.

$1052.00 - $1473.00

Specifications

Certifications/Compliance Certified RNase and DNase Free. ANSI Compliant.
Surface Treatment Non-Treated
Lid Without Lid
No. per Case 100
No. per Pack 25
View More Specs

Products 3
Catalog Number Mfr. No. Color Sterility Volume (Metric) Well Price Quantity & Availability  
Catalog Number Mfr. No. Color Sterility Volume (Metric) Well Price Quantity & Availability  
12-566-211
View Documents
Thermo Scientific™
267460
Natural Sterile 55 μL Case of 100 for $1,473.00
Add to cart
 
12-566-212
View Documents
Thermo Scientific™
267461
Black Non-sterile Case of 100 for $1,184.00
Add to cart
 
12-566-213
View Documents
Thermo Scientific™
267462
White Non-sterile 55 μL Case of 100 for $1,052.00
Add to cart
 
Description

Description

Lower spending when storing compounds for small volume applications by using Thermo Scientific™ Nunc™ 384-Shallow Well Standard Height Polypropylene Microplates. Polypropylene is ideal for storage plates because it has a lower binding capacity to prevent proteins or DNA binding, and allows for complete sample recovery. Withstands temperatures from -80° to +121°C. Offered in a variety of colors to allow for quick identification of storage plates.

Features:
  • Rounded wells are optimized for mixing and sample recovery
  • Square rounded wells minimize wicking (capillary action)
  • Natural plates for storage identification or storing colorimetric assays
  • White plates for luminescent assays
  • Black plates for fluorescent assays
  • Customized bar coding is available
Warranty: 90 days
Specifications

Specifications

Certified RNase and DNase Free. ANSI Compliant.
Without Lid
25
Non-Treated
100
384

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit