Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces
Reduce the volume of reagent needed with Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces.
Manufacturer: Thermo Scientific™ 164555/PRC
 View more versions of this product
Catalog No. 12-568-50
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Lid No
    Description Treated; no lid; Sterile; 25/100
    No. per Case 100
    No. per Pack 25
    Sterile Yes
    Surface Coating Cell Culture
    Volume (Metric) Total 120µL
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Well Design 384F
    Closure None,None
    Color Clear
    Well Shape Flat bottom
    Reduce the volume of reagent needed with Thermo Scientific™ Nunc™ 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces, which are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample volume reduction, providing cost savings and allowing more samples to be screened.
    Features:
    • Clear Polystyrene has excellent optical properties, making it ideal for optical measurements
    • Untreated Polystyrene is hydrophobic and suited for growth of suspension cultures that can proliferate and grow without attachment
    • Untreated surfaces ideal for adding coatings in the lab
    • Cell culture treated, hydrophilic surface promotes cell attachment and growth
    • Customized bar coding is available
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback