Thermo Scientific™ Nunc™ 384-Well Optical Bottom Plates Available on GSA/VA Contract for Federal Government customers only.
View microscopic plate contents using luminescence and fluorescence with these 384-well optical bottom plates.
Manufacturer: Thermo Scientific™ 240074
 View more versions of this product
Catalog No. 12-566-281
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Lid No
    No. per Case 30
    No. per Pack 5
    Sterile No
    Surface Coating Non-Treated
    Volume (Metric) Total 120µL
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Well Design 384 F
    Closure No,No
    Color Black
    Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Thermo Scientific™ Nunc™ MicroWell™ 384-Well Optical-Bottom Plates with Polymer or Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
    Features:
    • Poly- D-Lysine and Collagen coating creates a positive charge on the surface that ensures cell attachment, growth and differentiation.
    • Cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
    • Untreated Polystyrene is hydrophobic and suited for growth of suspension cultures that can proliferate and grow without attachment
    • Polystyrene upper structureis available with either a #1.5 cover glass or polystyrene base
    • Customized bar coding is available
  • Description & Specifications
Related Products
Product Content Feedback