Learn More
Thermo Scientific™ Nunc™ 384-Well Optical Bottom Plates Available on GSA/VA Contract for Federal Government customers only.
View microscopic plate contents using luminescence and fluorescence with these 384-well optical bottom plates.
Manufacturer:  Thermo Scientific™ 164730
 View more versions of this product
Catalog No. 12-565-473
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with Thermo Scientific™ Nunc™ MicroWell™ 384-Well Optical-Bottom Plates with Polymer or Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
  • Poly- D-Lysine and Collagen coating creates a positive charge on the surface that ensures cell attachment, growth and differentiation.
  • Cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Untreated Polystyrene is hydrophobic and suited for growth of suspension cultures that can proliferate and grow without attachment
  • Polystyrene upper structureis available with either a #1.5 cover glass or polystyrene base
  • Customized bar coding is available
Description & Specifications
Specifications
No
No
30
Yes
120µL
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Black
10
Cell Culture
384 F