Learn More
Thermo Scientific™ Nunc™ 384 Well ShallowWell Standard Height Black and White Plates Available on GSA/VA Contract for Federal Government customers only.
Save costs in samples and reagents by reducing the total well volume to 25µL using Nunc 384-well shallow well standard height black and white polystyrene plates.
Manufacturer:  Thermo Scientific™ 264705
 View more versions of this product
Catalog No. 12-565-746
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Save costs in samples and reagents by reducing the total well volume to 25µL using Thermo Scientific™ Nunc™ 384-well ShallowWell Standard Height Black and White Plates. The standard height of the plate allows you to use automation without making adjustments to robotics, readers or liquid handlers.
Background, autofluorescence and crosstalk are improved when you use Nunc black or white microplates.
  • Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
Description & Specifications
Specifications
Black
25
Non-Treated
384 U
100
No
25µL