Thermo Scientific™ Nunc™ 96-Well Polypropylene MicroWell™ Plates Available on GSA/VA Contract for Federal Government customers only.
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA to bind and allow for complete sample recovery.
Manufacturer: Thermo Scientific™ 267369
 View more versions of this product
Catalog No. 12-565-372
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Description Polypropylene microplates with shared-wall technology which increases well volume and widens cells to improve mixing
    No. per Case 120
    No. per Pack 10,10
    Sterile No
    Surface Coating Non-Treated
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Volume (Metric) Well 500µL,500µL
    Well Design U-bottom
    Color Red
    Item Description 96 Round Well, Red Polypropylene, Non-Treated, without lid, Non-sterile
    Achieve complete sample recovery with Thermo Scientific™ Nunc™ 96-Well Polypropylene MicroWell™ Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
    • Shared wall design increase the well volume
    • Wider wells and rounded bottom improve mixing
    • Conical bottom well are for optimal sample recovery
    • Excellent chemical resistance
    • Withstand temperatures from -80° to +121°C (-112° to +250°F)
    • Black and white plates are available for fluorescent- and luminescent-based applications
    • Low binding polypropylene for homogeneous assays and storage
  • Description & Specifications
Related Products
Product Content Feedback