Learn More
Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
Supplier: Thermo Scientific™ 267407
Description
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
- Shared-wall design increases the well volume
- Wider wells and rounded bottom improve mixing
- Conical bottom well for optimal sample recovery
- Black and white plates available for fluorescent- and luminescent-based applications
- Low binding polypropylene for homogeneous assays and storage
Specifications
No Barcode | |
96-well | |
96-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Yellow | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
500 μL | |
450 μL | |
Non-treated | |
Yes | |
120 | |
120/Cs. | |
Yes | |
ANSI/ SLAS Standard |
Certified RNase and DNase Free. ANSI Compliant. | |
Without Lid | |
8 x 12 | |
10 pk., 120 cs. | |
Nunc | |
96-well plate | |
Polypropylene | |
96 | |
Non-sterile | |
Round | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
10 | |
For Research Use Only. Not for use in diagnostic procedures. | |
Certified Quality |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.