Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ 96-Well Polypropylene Storage Microplates

Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and allow for complete sample recovery.

Manufacturer:  Thermo Scientific™ 95040410

 View more versions of this product

Catalog No. 14-387-92

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Achieve complete sample recovery with Thermo Scientific™ Nunc™ 96-Well Polypropylene MicroWell™ Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.

  • Shared wall design increase the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well are for optimal sample recovery
  • Excellent chemical resistance
  • Withstand temperatures from -80° to +121°C (-112° to +250°F)
  • Black and white plates are available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
Specifications

Specifications

V
Natural
Polypropylene
25
Square
500 μL
96
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Polypropylene microplates with shared-wall technology which increases well volume and widens cells to improve mixing
50
Non-sterile
Non-Treated
Standard
8 x 12
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.