Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Perform automated applications more precisely with these round-bottom plates, which are available with and without cell culture treatment.
Manufacturer: Thermo Scientific™ 174908
 View more versions of this product
Catalog No. 12-566-334
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Lid Yes
    No. per Case 8
    No. per Pack 8
    Sterile Yes
    Surface Coating HydroCell
    Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
    Volume (Metric) Well 300µL
    Well Design 96 U,96 U
    Closure Yes
    Color Clear
    Item Description 174908
    Perform automated applications more precisely with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates, which are available with and without cell culture treatment. The plates are optimized for robotics, automated readers and liquid handling systems, and are ideal for agglutination assays.
    • Clear polystyrene has excellent optical properties making it ideal for optical measurements
    • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
    • Untreated surfaces are ideal for adding coatings in your lab
    • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
    • Plates are optimized for robotics, plate readers and liquid handlers
    • Customized bar coding is available
  • Description & Specifications
Related Products
Product Content Feedback