Learn More
Thermo Scientific™ Nunc™ ArrayCote™ 16-Well Slides
Includes: Include lids and tool for removal of the upper structure.
Products 3
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
12-565-537
View Documents
Thermo Scientific™
277263
N/A 16-Well Glass Slide Aminopropylsilane This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
12-565-538
View Documents
Thermo Scientific™
277264
N/A 16-Well Glass Slide aldehyde This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
12-565-539
View Documents
Thermo Scientific™
277265
N/A 16-Well Glass Slide epoxy This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Standard 1 x 3 in. glass microarray slides with 16-well polystyrene upper stucture allow the analysis of multiple samples or conditions on the same slide. The upper structure can be detached, so the slide can be scanned like a standard microarray slide. A choice of surface coatings is available to suit a variety of applications. The aminopropylsilane surface permits attachment through nonspecific electrostatic adsorption; the aldehyde and epoxy surfaces enable covalent binding of amine-modified DNA. All three surfaces also permit protein binding.
All slides offer a color-coded frosted writing area for easy identification.
Order Info
Well dimensions are the same as Nunc ArrayCote 96-well Plates, allowing easy scale-up. Working volume: 100–200µL.