Learn More
Thermo Scientific™ Nunc™ Barcode Scanners, USB
Track samples simply using these bar code scanners. Plug and Play function can automatically switch to read codes: 128A, 128B, 128C, 39 and Interleve 2/5.
$1028.00
Products 1
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
12-565-001
View Documents
Thermo Scientific™
330050
any USB port Case for $1,028.00
Add to cart
Description
Description
Track samples simply using Thermo Scientific™ Nunc™ Barcode Scanners. “Plug and Play” function can automatically switch to read codes: 128A, 128B, 128C, 39 and Interleve 2/5. Convenient and easy to use with an Excel spread sheet.
Durable, low cost, and easy to use–simply plug into a USB or keyboard port.
  • Scanners automatically switch between Code 128, Code 39, Interleave 2 of 5 and several other barcodes
  • Must be in contact with the barcode to read
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.