Learn More
Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps Available on GSA/VA Contract for Federal Government customers only.
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific? Nunc? Cell Culture Treated Flasks with Filter Caps.
Manufacturer:  Thermo Scientific™ 178883
 View more versions of this product
Catalog No. 12-565-30
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Get growth areas from 25-175cm2 with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Filter Caps. These caps receive the Thermo Scientific™ Nunclon™ Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types. They are individually leak tested and come with filter caps allow constant airflow and minimize chance of contamination.
Nunc Flasks with Nunclon Delta treated surface are designed for enhanced cell attachment, growth and differentiation. Unique hydrophobic 0.22µm filter cap utilizes a three-dimensional matrix of nonwettable polyethylene fibers that prevent penetration of microorganisms.
  • Nunclon Delta Certification including testing on 4 different cell types is provided in each case
  • Short, wide necks allow for easy access
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
  • Barcoded 175cm2 flasks with large Code 128 are compatible with automation
Ordering Information:
Extra caps in each case.
Includes:
Caps
Description & Specifications
Specifications
No
175cm²
32
Nunclon? Delta
68mL
Filter
Straight
2
68mL