Thermo Scientific™ Nunc™ Cell Factory™ Hand Manipulator Systems
Automate the filling and emptying of medium or cell suspension into 4 x 3 Thermo Scientific™ Nunc™ Cell Factory 10 or 4 x 1 Nunc Cell Factory 40 systems.
Manufacturer: Thermo Scientific™ 176953
 View more versions of this product
Catalog No. 12-565-46
  Check Availability
Add to Cart
    Specifications
    Material Stainless Steel
    Description Cell Factory 40 Hand Manipulator
    No. per Pack 1
    No. per Case 1
    Thermo Scientific™ Nunc™ Cell Factory™ Hand Manipulators are designed for the safe and efficient handling of Cell Factory 10 and 40 layer systems. The Cell Factory Hand Manipulator is mobile and includes a foot brake for safe placement. The Cell Factory 10 Hand Manipulator is compact and ideal for use within in a biosafety cabinet.
    • Constructed of stainless steel, designed for use in clean room environments
  • Description & Specifications
Related Products
Product Content Feedback