Thermo Scientific™ Nunc™ Cell Factory™ System Accessories Available on GSA/VA Contract for Federal Government customers only.
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Manufacturer: Thermo Scientific™ 173248
 View more versions of this product
Catalog No. 12-565-109
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With Cell Factory Systems (all models)
    Description Airvent Filter with HDPE Connector, 1µm
    No. per Pack 2
    No. per Case 2
    Pore Size (Metric) 1µm
    Sterile Yes
    Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
    Connections Made Easy:
    • Plug a media bag or tubing set to any Cell Factory system for enhanced aseptic connectivity for filling and harvesting
    Additional Features:
    • Save time and reduce costs
    • Minimize cleaning processes such as autoclaving and associated validations
    • Prevent build up of back pressure during filling application
    • Get additional air venting with the venting options
  • Description & Specifications
Related Products
Product Content Feedback